missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine-2-carboxylic acid, 97%

CAS: 1462-86-8 | C6H6N2O2 | 138.126 g/mol

$126.05 - $443.10

Chemical Identifiers

CAS 1462-86-8
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00090153
InChI Key BOOMHTFCWOJWFO-UHFFFAOYSA-N
Synonym 3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
PubChem CID 73836
IUPAC Name 3-aminopyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6486106
View Documents
Thermo Scientific Chemicals
H6486106
5 g
Each for $126.05
Only null left
 
AAH6486114
View Documents
Thermo Scientific Chemicals
H6486114
25 g
Each for $443.10
Only null left
 
Description

Description

Employed as an important intermediate for syntheses material and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for syntheses material and pharmaceutical.

Solubility
Soluble in acetone, ethanol and methanol.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1462-86-8
138.126
BOOMHTFCWOJWFO-UHFFFAOYSA-N
73836
C1=CC(=C(N=C1)C(=O)O)N
C6H6N2O2
MFCD00090153
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
3-aminopyridine-2-carboxylic acid

Specifications

1462-86-8
Brown
MFCD00090153
3600
Soluble in acetone,ethanol and methanol.
C1=CC(=C(N=C1)C(=O)O)N
138.126
138.13
Solid
218°C to 222°C
C6H6N2O2
5 g
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopyridine-2-carboxylic acid
73836
98%
3-Aminopyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

EINECSNumber : 215-971-8

RTECSNumber : US5175000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only