missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridazine 98.0+%, TCI America™

Catalog No. p-7117698
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20481G 1 g
A20485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20481G Supplier TCI America Supplier No. A20481G
Only null left

Chemical Identifiers

CAS 5469-70-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD01529869
InChI Key LETVJWLLIMJADE-UHFFFAOYSA-N
Synonym 3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin
PubChem CID 230373
IUPAC Name pyridazin-3-amine
SMILES C1=CC(=NN=C1)N

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 1 g
Formula Weight 95.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyridazine