accessibility menu, dialog, popup

UserName

CAS RN 5469-70-5

IUPAC Name: pyridazin-3-amine
Synonyms: pyridazin-3-amine
Molecular Weight (g/mol): 95.11
Molecular Formula: C4H5N3
InChi Key: LETVJWLLIMJADE-UHFFFAOYSA-N
SMILES: NC1=CC=CN=N1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)