missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazole 98.0+%, TCI America™

Catalog No. p-7117610
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18595G 5 g
A185925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18595G Supplier TCI America Supplier No. A18595G
Only null left

Chemical Identifiers

CAS 1820-80-0
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.094
InChI Key JVVRJMXHNUAPHW-UHFFFAOYSA-N
Synonym 3-aminopyrazole, 1h-pyrazol-3-amine, 1h-pyrazole-5-amine, 5-aminopyrazole, 1h-pyrazol-3-ylamine, 3-amino-1h-pyrazole, 3-aminopyrozole, 5-amino-1h-pyrazole, aminopyrazole, 3-pyrazolamine
PubChem CID 74561
IUPAC Name 1H-pyrazol-5-amine
SMILES C1=C(NN=C1)N

Specifications

Color White-Yellow
Boiling Point 282°C
Quantity 5 g
Formula Weight 83.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Aminopyrazole
TSCA No
Recommended Storage Refrigerator