accessibility menu, dialog, popup

UserName

CAS RN 1820-80-0

IUPAC Name: 1H-pyrazol-5-amine
Synonyms: 2H-pyrazol-3-amine
Molecular Weight (g/mol): 83.09
Molecular Formula: C3H5N3
InChi Key: JVVRJMXHNUAPHW-UHFFFAOYSA-N
SMILES: NC1=CC=NN1
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)