missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopropyldiethoxymethylsilane 97.0+%, TCI America™
Supplier: TCI America D1980500ML
Specifications
| 3-Aminopropyldiethoxymethylsilane | |
| Yellow | |
| 88°C | |
| MFCD00039785 | |
| 3267 | |
| HXLAEGYMDGUSBD-UHFFFAOYSA-N | |
| 3-[diethoxy(methyl)silyl]propan-1-amine | |
| 18511 | |
| ≥97.0% (GC,T) |
| 3179-76-8 | |
| 11 (20g/L H2O soln. 20°C) | |
| C8H21NO2Si | |
| 500 mL | |
| 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane | |
| CCO[Si](C)(CCCN)OCC | |
| 191.346 | |
| 191.35 | |
| Liquid |
Chemical Identifiers
| 3179-76-8 | |
| 191.346 | |
| HXLAEGYMDGUSBD-UHFFFAOYSA-N | |
| 18511 | |
| CCO[Si](C)(CCCN)OCC |
| C8H21NO2Si | |
| MFCD00039785 | |
| 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane | |
| 3-[diethoxy(methyl)silyl]propan-1-amine |
Safety and Handling
RTECSNumber : UI0700000
TSCA : Yes