missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyldiethoxymethylsilane 97.0+%, TCI America™

Catalog No. p-7127046
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D198025ML 25 mL
D1980100ML 100 mL
D1980500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D198025ML Supplier TCI America Supplier No. D198025ML
Only null left

Chemical Identifiers

CAS 3179-76-8
Molecular Formula C8H21NO2Si
Molecular Weight (g/mol) 191.346
MDL Number MFCD00039785
InChI Key HXLAEGYMDGUSBD-UHFFFAOYSA-N
Synonym 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane
PubChem CID 18511
IUPAC Name 3-[diethoxy(methyl)silyl]propan-1-amine
SMILES CCO[Si](C)(CCCN)OCC

Specifications

Color Yellow
pH 11 (20g/L H2O soln. 20°C)
Boiling Point 88°C
Quantity 25 mL
UN Number 3267
Formula Weight 191.35
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminopropyldiethoxymethylsilane
RTECSNumber UI0700000
TSCA Yes