missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile Fumarate 98.0+%, TCI America™

$178.95 - $1,625.46

Chemical Identifiers

CAS 2079-89-2
Molecular Formula C3H6N2
MDL Number MFCD00078321
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A079625G
View Documents
TCI America
A079625G
25 g N/A
Only null left
 
A0796500G
View Documents
TCI America
A0796500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2079-89-2
C3H6N2
MFCD00078321

Specifications

2079-89-2
C3H6N2
25 g
≥98.0% (T)
3-Aminopropionitrile Fumarate
White-Yellow
MFCD00078321
256.26
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UG0700000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'