missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile Fumarate 98.0+%, TCI America™

Catalog No. A0796500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0796500G 500 g
A079625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0796500G Supplier TCI America Supplier No. A0796500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2079-89-2
Molecular Formula C3H6N2
MDL Number MFCD00078321

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 256.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopropionitrile Fumarate
RTECSNumber UG0700000
TSCA Yes