missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile, 98%, stab. with potassium carbonate

Catalog No. AAA1304303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1304303 1 g
AAA1304306 5 g
AAA1304314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1304303 Supplier Thermo Scientific Chemicals Supplier No. A1304303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 151-18-8 | C3H6N2 | 70.10 g/mol

3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

Solubility
Soluble in water.

Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.

Chemical Identifiers

CAS 151-18-8
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.10
MDL Number MFCD00014820
InChI Key AGSPXMVUFBBBMO-UHFFFAOYSA-N
Synonym 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino
PubChem CID 1647
ChEBI CHEBI:27413
IUPAC Name 3-aminopropanenitrile
SMILES NCCC#N

Specifications

Density 0.952
Boiling Point 79°C to 81°C (16 mmHg)
Odor Amine-like
Refractive Index 1.4375
Linear Formula H2NCH2CH2CN
Quantity 1 g
UN Number UN2735
Beilstein 1698848
Sensitivity Air and moisture sensitive
Merck Index 14,469
Solubility Information Soluble in water.
Formula Weight 70.1
Percent Purity 98%
Chemical Name or Material 3-Aminopropionitrile, stabilized with potassium carbonate
Show More Show Less
Hazard Category H227-H302-H314-H335-H361
Hazard Statement GHS H Statement
H301-H314-H318-H361-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 205-786-0
RTECSNumber UG0350000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only