accessibility menu, dialog, popup

UserName

CAS RN 151-18-8

IUPAC Name: 3-aminopropanenitrile
Synonyms: β aminopropionitrile bapn fumarate β-alaninenitrile lathyrus protein lathyrus poison lathyrus poison (substance) BAPN 3-aminopropiononitrile β-alaminenitrile
Molecular Weight (g/mol): 70.1
Molecular Formula: C3H6N2
InChi Key: AGSPXMVUFBBBMO-UHFFFAOYSA-N
SMILES: NCCC#N
Boiling Point 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)