missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminophthalic Acid 95.0+%, TCI America™
$85.75 - $247.92
Chemical Identifiers
| CAS | 5434-20-8 |
|---|---|
| Molecular Formula | C8H7NO4 |
| Molecular Weight (g/mol) | 181.147 |
| InChI Key | WGLQHUKCXBXUDV-UHFFFAOYSA-N |
| Synonym | 3-amino-1,2-benzenedicarboxylic acid, phthalic acid, 3-amino, 3-aminophthalicacid, 1,2-benzenedicarboxylic acid, 3-amino, 3-amino-phthalic acid, unii-7xv0v19zdg, 3-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, amino, 7xv0v19zdg, aminophthalic acid |
| PubChem CID | 79490 |
| IUPAC Name | 3-aminophthalic acid |
| SMILES | C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O |
Chemical Identifiers
| 5434-20-8 | |
| 181.147 | |
| 3-amino-1,2-benzenedicarboxylic acid, phthalic acid, 3-amino, 3-aminophthalicacid, 1,2-benzenedicarboxylic acid, 3-amino, 3-amino-phthalic acid, unii-7xv0v19zdg, 3-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, amino, 7xv0v19zdg, aminophthalic acid | |
| 3-aminophthalic acid |
| C8H7NO4 | |
| WGLQHUKCXBXUDV-UHFFFAOYSA-N | |
| 79490 | |
| C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O |
Specifications
| 5434-20-8 | |
| C8H7NO4 | |
| 3-amino-1,2-benzenedicarboxylic acid, phthalic acid, 3-amino, 3-aminophthalicacid, 1,2-benzenedicarboxylic acid, 3-amino, 3-amino-phthalic acid, unii-7xv0v19zdg, 3-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, amino, 7xv0v19zdg, aminophthalic acid | |
| C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O | |
| 181.147 | |
| 181.15 | |
| Crystalline Powder |
| 183°C | |
| 1 g | |
| WGLQHUKCXBXUDV-UHFFFAOYSA-N | |
| 3-aminophthalic acid | |
| 79490 | |
| ≥95.0% (T) | |
| 3-Aminophthalic Acid |
Safety and Handling
TSCA : Yes