missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic Acid Monohydrate (contains varying amounts of Anhydride), TCI America™

Catalog No. A12811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12811G Supplier TCI America Supplier No. A12811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 206658-89-1
Molecular Formula C6H10BNO3
Molecular Weight (g/mol) 154.96
MDL Number MFCD00149554
InChI Key XAEOVQODHLLNKX-UHFFFAOYSA-N
Synonym 3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate, boronic acid, 3-aminophenyl-, monohydrate, 3-aminobenzeneboronic acid hydrate, pubchem4016, acmc-1cfgg, 3-boronoaniline monohydrate
PubChem CID 14389423
IUPAC Name (3-aminophenyl)boronic acid hydrate
SMILES O.NC1=CC=CC(=C1)B(O)O

Specifications

Quantity 1 g
Formula Weight 136.95
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenylboronic Acid Monohydrate (contains varying amounts of Anhydride)