accessibility menu, dialog, popup

UserName

CAS RN 206658-89-1

IUPAC Name: (3-aminophenyl)boronic acid hydrate
Synonyms: M-aminophenylboronic acid hydrate
Molecular Weight (g/mol): 154.96
Molecular Formula: C6H10BNO3
InChi Key: XAEOVQODHLLNKX-UHFFFAOYSA-N
SMILES: O.NC1=CC=CC(=C1)B(O)O
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)