missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic acid monohydrate, 98%

Catalog No. p-3756821
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC352760010 1 g Glass bottle
AC352760050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC352760010 Supplier Thermo Scientific Chemicals Supplier No. 352760010
Only null left

CAS: 206658-89-1 | C6H10BNO3 | 154.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 206658-89-1
Molecular Formula C6H10BNO3
Molecular Weight (g/mol) 154.96
MDL Number MFCD00149554
InChI Key XAEOVQODHLLNKX-UHFFFAOYSA-N
Synonym 3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate, boronic acid, 3-aminophenyl-, monohydrate, 3-aminobenzeneboronic acid hydrate, pubchem4016, acmc-1cfgg, 3-boronoaniline monohydrate
PubChem CID 14389423
IUPAC Name (3-aminophenyl)boronic acid;hydrate
SMILES O.NC1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 94°C to 97°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2C6H4B(OH)2·H2O
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 154.97
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Aminophenylboronic acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only