missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylacetylene, 98%

Catalog No. AAL2012406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2012406 5 g
AAL2012414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2012406 Supplier Thermo Scientific Chemicals Supplier No. L2012406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54060-30-9 | C8H7N | 117.15 g/mol

It is used as Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.

Chemical Identifiers

CAS 54060-30-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00014779
InChI Key NNKQLUVBPJEUOR-UHFFFAOYSA-N
Synonym 3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
PubChem CID 104682
IUPAC Name 3-ethynylaniline
SMILES NC1=CC=CC(=C1)C#C

Specifications

Melting Point 22°C
Density 1.04
Boiling Point 70°C (1 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.617
Quantity 5 g
Beilstein 2935417
Solubility Information Insoluble in water.
Formula Weight 117.15
Percent Purity 98%
Chemical Name or Material 3-Aminophenylacetylene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 258-944-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only