missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%

Catalog No. p-7035566
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53284MD 250 mg
AAH5328403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53284MD Supplier Thermo Scientific Chemicals Supplier No. H53284MD
Only null left

CAS: 352525-94-1 | C7H11BClNO2 | 187.43 g/mol

3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.

Chemical Identifiers

CAS 352525-94-1
Molecular Formula C7H11BClNO2
Molecular Weight (g/mol) 187.43
MDL Number MFCD02093050
InChI Key NPTBTFRGCBFYPZ-UHFFFAOYSA-N
Synonym 3-aminomethyl phenyl boronic acid hydrochloride, 3-aminomethyl phenylboronic acid hydrochloride, 3-aminomethylphenylboronic acid hydrochloride, 3-aminomethylphenyl boronic acid hydrochloride, 3-aminomethylphenylboronic acid hcl, 3-aminomethylphenyl boronic acid hcl, boronic acid, 3-aminomethyl phenyl-, hydrochloride, 3-aminomethylphenylboronic acid, hcl
PubChem CID 16427085
IUPAC Name [3-(aminomethyl)phenyl]boronic acid;hydrochloride
SMILES B(C1=CC(=CC=C1)CN)(O)O.Cl

Specifications

Melting Point 248°C to 250°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 187.43
Percent Purity 96%
Chemical Name or Material 3-(Aminomethyl)benzeneboronic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only