missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobiphenyl 99.0+%, TCI America™
Supplier: TCI America B098725G
Specifications
| 3-Aminobiphenyl | |
| White-Yellow | |
| C12H11N | |
| 25 g | |
| MUNOBADFTHUUFG-UHFFFAOYSA-N | |
| [1,1'-biphenyl]-3-amine | |
| 16717 | |
| ≥99.0% (GC,T) |
| 2243-47-2 | |
| 254°C | |
| MFCD00047846 | |
| 3-Aminodiphenyl, 3-Biphenylamine | |
| NC1=CC=CC(=C1)C1=CC=CC=C1 | |
| 169.23 | |
| 169.23 | |
| Crystalline Lumps |
Chemical Identifiers
| 2243-47-2 | |
| 169.23 | |
| MUNOBADFTHUUFG-UHFFFAOYSA-N | |
| 16717 | |
| NC1=CC=CC(=C1)C1=CC=CC=C1 |
| C12H11N | |
| MFCD00047846 | |
| 3-Aminodiphenyl, 3-Biphenylamine | |
| [1,1'-biphenyl]-3-amine |
Safety and Handling
RTECSNumber : DU8900000
TSCA : No