CAS RN 2243-47-2
CAS RN 2243-47-2
IUPAC Name:
[1,1'-biphenyl]-3-amine
Synonyms:
[1,1'-biphenyl]-3-amine
Molecular Weight (g/mol):
169.23
Molecular Formula:
C12H11N
InChi Key:
MUNOBADFTHUUFG-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Aminobiphenyl 99.0+%, TCI America™
CAS: 2243-47-2 Molecular Formula: C12H11N Molecular Weight (g/mol): 169.23 MDL Number: MFCD00047846 InChI Key: MUNOBADFTHUUFG-UHFFFAOYSA-N Synonym: 3-Aminodiphenyl, 3-Biphenylamine PubChem CID: 16717 IUPAC Name: [1,1'-biphenyl]-3-amine SMILES: NC1=CC=CC(=C1)C1=CC=CC=C1