missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobiphenyl 99.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 254°C |
| Quantity | 5 g |
| Formula Weight | 169.23 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 3-Aminobiphenyl |
Safety and Handling
| RTECSNumber | DU8900000 |
| TSCA | No |