missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7117808
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22595G 5 g
A225925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22595G Supplier TCI America Supplier No. A22595G
Only null left

Chemical Identifiers

CAS 1877-77-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007817
InChI Key OJZQOQNSUZLSMV-UHFFFAOYSA-N
Synonym 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol
PubChem CID 80293
IUPAC Name (3-aminophenyl)methanol
SMILES NC1=CC=CC(CO)=C1

Specifications

Melting Point 94°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzyl Alcohol
RTECSNumber DN3154450
TSCA No