missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzyl alcohol, 98+%

Catalog No. AAL0784714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0784714 25 g
AAL0784706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0784714 Supplier Thermo Scientific Chemicals Supplier No. L0784714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1877-77-6 | C7H9NO | 123.16 g/mol

3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1877-77-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007817
InChI Key OJZQOQNSUZLSMV-UHFFFAOYSA-N
Synonym 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol
PubChem CID 80293
IUPAC Name (3-aminophenyl)methanol
SMILES NC1=CC=CC(CO)=C1

Specifications

Melting Point 91°C to 95°C
Quantity 25 g
Beilstein 2205844
Sensitivity Air and light sensitive
Solubility Information Soluble in water.
Formula Weight 123.16
Percent Purity ≥98%
Chemical Name or Material 3-Aminobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-514-8
RTECSNumber DN3154450
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only