Learn More
3-Aminobenzyl alcohol, 97%
CAS: 1877-77-6 | C7H9NO | 123.16 g/mol
$120.93 - $120.93
Chemical Identifiers
| CAS | 1877-77-6 |
|---|---|
| Molecular Formula | C7H9NO |
| Molecular Weight (g/mol) | 123.16 |
| MDL Number | MFCD00007817 |
| InChI Key | OJZQOQNSUZLSMV-UHFFFAOYSA-N |
| Synonym | 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol |
| PubChem CID | 80293 |
| IUPAC Name | (3-aminophenyl)methanol |
| SMILES | NC1=CC=CC(CO)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC184570050
|
Thermo Scientific Chemicals
184570050 |
5 g | Glass bottle |
Each for $120.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1877-77-6 | |
| 123.16 | |
| OJZQOQNSUZLSMV-UHFFFAOYSA-N | |
| 80293 | |
| NC1=CC=CC(CO)=C1 |
| C7H9NO | |
| MFCD00007817 | |
| 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol | |
| (3-aminophenyl)methanol |
Specifications
| 1877-77-6 | |
| 100.0 | |
| Beige or Beige-Gray to Brown | |
| 96% min. (GC) | |
| C7H9NO | |
| MFCD00007817 | |
| 13, 619 | |
| OJZQOQNSUZLSMV-UHFFFAOYSA-N | |
| (3-aminophenyl)methanol | |
| 80293 | |
| 97% | |
| 3-Aminobenzyl alcohol |
| 96.0 | |
| 91.0°C to 95.0°C | |
| Authentic | |
| Glass bottle | |
| H2NC6H4CH2OH | |
| 5 g | |
| 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol | |
| NC1=CC=CC(CO)=C1 | |
| 123.16 | |
| 123.15 | |
| Fine Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 217-514-8
RUO – Research Use Only