missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzonitrile 98.0+%, TCI America™

Supplier:  TCI America A08605G

 View more versions of this product

Catalog No. A08605G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Aminobenzonitrile
53°C
C7H6N2
5 g
3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
C1=CC(=CC(=C1)N)C#N
118.139
118.14
Crystalline Powder
2237-30-1
290°C
MFCD00007756
3439
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-aminobenzonitrile
16702
≥98.0% (GC,T)

Chemical Identifiers

2237-30-1
118.139
NJXPYZHXZZCTNI-UHFFFAOYSA-N
16702
C1=CC(=CC(=C1)N)C#N
C7H6N2
MFCD00007756
3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
3-aminobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1812

RTECSNumber : DI2454000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions