missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzonitrile 98.0+%, TCI America™

Catalog No. p-7117094
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08605G 5 g
A086025G 25 g
A0860250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A08605G Supplier TCI America Supplier No. A08605G
Only null left

Chemical Identifiers

CAS 2237-30-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00007756
InChI Key NJXPYZHXZZCTNI-UHFFFAOYSA-N
Synonym 3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
PubChem CID 16702
IUPAC Name 3-aminobenzonitrile
SMILES C1=CC(=CC(=C1)N)C#N

Specifications

Melting Point 53°C
Boiling Point 290°C
Quantity 5 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzonitrile
EINECSNumber (3)-1812
RTECSNumber DI2454000
TSCA No