Learn More
3-Aminobenzonitrile, 99%
CAS: 2237-30-1 | C7H6N2 | 118.139 g/mol
Supplier: Thermo Scientific Chemicals A1399518
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Aminobenzonitrile | |
| 51°C to 53°C | |
| >110°C (230°F) | |
| C7H6N2 | |
| 50 g | |
| 3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile | |
| C1=CC(=CC(=C1)N)C#N | |
| 118.139 | |
| 118.14 |
| 2237-30-1 | |
| 290°C | |
| Almond-like | |
| MFCD00007756 | |
| UN3439 | |
| NJXPYZHXZZCTNI-UHFFFAOYSA-N | |
| 3-aminobenzonitrile | |
| 16702 | |
| 99% |
Chemical Identifiers
| 2237-30-1 | |
| 118.139 | |
| NJXPYZHXZZCTNI-UHFFFAOYSA-N | |
| 16702 | |
| C1=CC(=CC(=C1)N)C#N |
| C7H6N2 | |
| MFCD00007756 | |
| 3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile | |
| 3-aminobenzonitrile |
Safety and Handling
GHS H Statement
H311-H302-H317
Toxic in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H315-H317-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 218-800-5
RTECSNumber : DI2454000
TSCA : No
RUO – Research Use Only