missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzonitrile, 99%

Catalog No. p-7021595
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1399509 10 g
AAA1399518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1399509 Supplier Thermo Scientific Chemicals Supplier No. A1399509
Only null left

CAS: 2237-30-1 | C7H6N2 | 118.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2237-30-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00007756
InChI Key NJXPYZHXZZCTNI-UHFFFAOYSA-N
Synonym 3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
PubChem CID 16702
IUPAC Name 3-aminobenzonitrile
SMILES C1=CC(=CC(=C1)N)C#N

Specifications

Melting Point 51°C to 53°C
Boiling Point 290°C
Flash Point >110°C (230°F)
Odor Almond-like
Quantity 10 g
UN Number UN3439
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 3-Aminobenzonitrile
Hazard Category H301+H311+H331-H315-H317-H319
Hazard Statement GHS H Statement
H311-H302-H317
Toxic in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 218-800-5
RTECSNumber DI2454000
TSCA No

RUO – Research Use Only