missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzoic Acid 99.0+%, TCI America™
$329.94 - $2031.83
Chemical Identifiers
| CAS | 99-05-8 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.138 |
| MDL Number | MFCD00007795 |
| InChI Key | XFDUHJPVQKIXHO-UHFFFAOYSA-N |
| Synonym | m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u |
| PubChem CID | 7419 |
| ChEBI | CHEBI:42682 |
| IUPAC Name | 3-aminobenzoic acid |
| SMILES | C1=CC(=CC(=C1)N)C(=O)O |
Chemical Identifiers
| 99-05-8 | |
| 137.138 | |
| XFDUHJPVQKIXHO-UHFFFAOYSA-N | |
| 7419 | |
| 3-aminobenzoic acid |
| C7H7NO2 | |
| MFCD00007795 | |
| m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u | |
| CHEBI:42682 | |
| C1=CC(=CC(=C1)N)C(=O)O |
Specifications
| 99-05-8 | |
| White-Yellow | |
| MFCD00007795 | |
| m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u | |
| C1=CC(=CC(=C1)N)C(=O)O | |
| 137.138 | |
| CHEBI:42682 | |
| ≥99.0% (T) | |
| 3-Aminobenzoic Acid |
| 174°C | |
| C7H7NO2 | |
| 25 g | |
| XFDUHJPVQKIXHO-UHFFFAOYSA-N | |
| 3-aminobenzoic acid | |
| 7419 | |
| 137.14 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1454
RTECSNumber : DG1225000
TSCA : Yes