missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic Acid 99.0+%, TCI America™

Catalog No. A026825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A026825G 25 g
A0268500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A026825G Supplier TCI America Supplier No. A026825G

Chemical Identifiers

CAS 99-05-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007795
InChI Key XFDUHJPVQKIXHO-UHFFFAOYSA-N
Synonym m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u
PubChem CID 7419
ChEBI CHEBI:42682
IUPAC Name 3-aminobenzoic acid
SMILES C1=CC(=CC(=C1)N)C(=O)O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzoic Acid
EINECSNumber (3)-1454
RTECSNumber DG1225000
TSCA Yes