missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic acid, 98%

Catalog No. aaa195400e
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA195400E 2500 g
AAA1954022 100 g
AAA1954036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA195400E Supplier Thermo Scientific Chemicals Supplier No. A195400E
Only null left

CAS: 99-05-8 | C7H7NO2 | 137.138 g/mol

It finds its application as crystalline Intermediate for azo dyes, pharmaceuticals, and X-ray contrast media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as crystalline Intermediate for azo dyes, pharmaceuticals, and X-ray contrast media.

Solubility
Soluble in water at 20°C 5.9g/L. Solubility in hot methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 99-05-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007795
InChI Key XFDUHJPVQKIXHO-UHFFFAOYSA-N
Synonym m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u
PubChem CID 7419
ChEBI CHEBI:42682
IUPAC Name 3-aminobenzoic acid
SMILES C1=CC(=CC(=C1)N)C(=O)O

Specifications

Melting Point 172°C to 179°C
Density 1.51
Flash Point 150°C (302°F)
Quantity 2500 g
Beilstein 471603
Merck Index 14,421
Solubility Information Soluble in water at 20°C 5.9g/L. Solubility in hot methanol is almost transparent.
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material 3-Aminobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-724-4
RTECSNumber DG1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only