missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzenesulfonamide 98.0+%, TCI America™
$171.34 - $171.34
Chemical Identifiers
| CAS | 98-18-0 |
|---|---|
| Molecular Formula | C6H8N2O2S |
| Molecular Weight (g/mol) | 172.202 |
| MDL Number | MFCD00035781 |
| InChI Key | JPVKCHIPRSQDKL-UHFFFAOYSA-N |
| Synonym | metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide |
| PubChem CID | 7377 |
| IUPAC Name | 3-aminobenzenesulfonamide |
| SMILES | C1=CC(=CC(=C1)S(=O)(=O)N)N |
Chemical Identifiers
| 98-18-0 | |
| 172.202 | |
| JPVKCHIPRSQDKL-UHFFFAOYSA-N | |
| 7377 | |
| C1=CC(=CC(=C1)S(=O)(=O)N)N |
| C6H8N2O2S | |
| MFCD00035781 | |
| metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide | |
| 3-aminobenzenesulfonamide |
Specifications
| 98-18-0 | |
| C6H8N2O2S | |
| 25 g | |
| JPVKCHIPRSQDKL-UHFFFAOYSA-N | |
| 3-aminobenzenesulfonamide | |
| 7377 | |
| ≥98.0% (HPLC,T) | |
| 3-Aminobenzenesulfonamide |
| 142°C | |
| MFCD00035781 | |
| metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide | |
| C1=CC(=CC(=C1)S(=O)(=O)N)N | |
| 172.202 | |
| 172.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2179
RTECSNumber : OY2200000
TSCA : Yes