missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid, 98%

Catalog No. p-7035195
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5290906 5 g
AAH5290914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5290906 Supplier Thermo Scientific Chemicals Supplier No. H5290906
Only null left

CAS: 30418-59-8 | C6H8BNO2 | 136.945 g/mol

3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

Solubility
Soluble in water (partly miscible).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents and heat.

Chemical Identifiers

CAS 30418-59-8
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD00007755
InChI Key JMZFEHDNIAQMNB-UHFFFAOYSA-N
Synonym 3-aminobenzeneboronic acid, 3-aminophenyl boronic acid, m-aminophenylboronic acid, 3-aminophenyl boranediol, m-aminophenyl boronic acid, 3-amino phenylboronic acid, chembl20852, boronic acid, 3-aminophenyl, m-aminophenyl metaboric acid
PubChem CID 92269
IUPAC Name (3-aminophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)N)(O)O

Specifications

Melting Point 138°C to 143°C
Quantity 5 g
Solubility Information Soluble in water (partly miscible).
Formula Weight 136.95
Percent Purity 98%
Chemical Name or Material 3-Aminobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 250-189-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only