accessibility menu, dialog, popup

UserName

NH2BOHHO

CAS RN 30418-59-8

IUPAC Name: (3-aminophenyl)boronic acid
Synonyms: M-aminophenylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: JMZFEHDNIAQMNB-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results