missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzamide 98.0+%, TCI America™
$67.11 - $227.41
Chemical Identifiers
| CAS | 3544-24-9 |
|---|---|
| Molecular Formula | C7H8N2O |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00007989 |
| InChI Key | GSCPDZHWVNUUFI-UHFFFAOYSA-N |
| Synonym | m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide |
| PubChem CID | 1645 |
| ChEBI | CHEBI:64042 |
| IUPAC Name | 3-aminobenzamide |
| SMILES | NC(=O)C1=CC=CC(N)=C1 |
Chemical Identifiers
| 3544-24-9 | |
| 136.15 | |
| GSCPDZHWVNUUFI-UHFFFAOYSA-N | |
| 1645 | |
| 3-aminobenzamide |
| C7H8N2O | |
| MFCD00007989 | |
| m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide | |
| CHEBI:64042 | |
| NC(=O)C1=CC=CC(N)=C1 |
Specifications
| 3544-24-9 | |
| White-Yellow | |
| MFCD00007989 | |
| m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide | |
| NC(=O)C1=CC=CC(N)=C1 | |
| 136.15 | |
| CHEBI:64042 | |
| ≥98.0% (T) | |
| 3-Aminobenzamide |
| 114°C | |
| C7H8N2O | |
| 5 g | |
| GSCPDZHWVNUUFI-UHFFFAOYSA-N | |
| 3-aminobenzamide | |
| 1645 | |
| 136.15 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CU8992000
TSCA : No