missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzamide 98.0+%, TCI America™
Supplier: TCI America A06305G
Specifications
| 3-Aminobenzamide | |
| 114°C | |
| C7H8N2O | |
| 5 g | |
| GSCPDZHWVNUUFI-UHFFFAOYSA-N | |
| 3-aminobenzamide | |
| 1645 | |
| 136.15 | |
| Crystalline Powder |
| 3544-24-9 | |
| White-Yellow | |
| MFCD00007989 | |
| m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide | |
| NC(=O)C1=CC=CC(N)=C1 | |
| 136.15 | |
| CHEBI:64042 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3544-24-9 | |
| 136.15 | |
| GSCPDZHWVNUUFI-UHFFFAOYSA-N | |
| 1645 | |
| 3-aminobenzamide |
| C7H8N2O | |
| MFCD00007989 | |
| m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide | |
| CHEBI:64042 | |
| NC(=O)C1=CC=CC(N)=C1 |
Safety and Handling
RTECSNumber : CU8992000
TSCA : No