missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-9-ethylcarbazole 90.0+%, TCI America™

Catalog No. A15085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15085G Supplier TCI America Supplier No. A15085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132-32-1
Molecular Formula C14H14N2
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004964
InChI Key OXEUETBFKVCRNP-UHFFFAOYSA-N
Synonym 3-amino-9-ethylcarbazole, 9-ethyl-9h-carbazol-3-amine, 3-amino-n-ethylcarbazole, 9h-carbazol-3-amine, 9-ethyl, carbazole, 3-amino-9-ethyl, 9-ethylcarbazol-3-ylamine, unii-8q2bg27jbu, 9-ethylcarbazole-3-ylamine, 8q2bg27jbu, 9-ethyl-9h-carbazol-3-ylamine
PubChem CID 8588
IUPAC Name 9-ethylcarbazol-3-amine
SMILES CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31

Specifications

Melting Point 103°C
Quantity 5 g
UN Number 2811
Formula Weight 210.28
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-9-ethylcarbazole
EINECSNumber (5)-0153
RTECSNumber FE3590000
TSCA Yes