accessibility menu, dialog, popup

UserName

CAS RN 132-32-1

IUPAC Name: 9-ethyl-9H-carbazol-3-amine
Synonyms: 3-amino-9-ethylcarbazole
Molecular Weight (g/mol): 210.28
Molecular Formula: C14H14N2
InChi Key: OXEUETBFKVCRNP-UHFFFAOYSA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(N)=C2
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)