missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-6-chloropyridazine 98.0+%, TCI America™
Supplier: TCI America A181725G
Specifications
| 3-Amino-6-chloropyridazine | |
| C4H4ClN3 | |
| 25 g | |
| DTXVKPOKPFWSFF-UHFFFAOYSA-N | |
| 6-chloropyridazin-3-amine | |
| 21643 | |
| ≥98.0% (GC,T) |
| 5469-69-2 | |
| MFCD00051506 | |
| 3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine | |
| NC1=CC=C(Cl)N=N1 | |
| 129.55 | |
| 129.55 | |
| Crystalline Powder |
Chemical Identifiers
| 5469-69-2 | |
| 129.55 | |
| DTXVKPOKPFWSFF-UHFFFAOYSA-N | |
| 21643 | |
| NC1=CC=C(Cl)N=N1 |
| C4H4ClN3 | |
| MFCD00051506 | |
| 3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine | |
| 6-chloropyridazin-3-amine |
Safety and Handling
TSCA : Yes