missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-chloropyridazine 98.0+%, TCI America™

Catalog No. p-7117593
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18175G 5 g
A181725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18175G Supplier TCI America Supplier No. A18175G
Only null left

Chemical Identifiers

CAS 5469-69-2
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00051506
InChI Key DTXVKPOKPFWSFF-UHFFFAOYSA-N
Synonym 3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
PubChem CID 21643
IUPAC Name 6-chloropyridazin-3-amine
SMILES NC1=CC=C(Cl)N=N1

Specifications

Quantity 5 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-6-chloropyridazine