missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(trifluoromethyl)benzoic acid, 97%

Catalog No. AAH2616906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2616906 5 g
AAH2616903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2616906 Supplier Thermo Scientific Chemicals Supplier No. H2616906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 328-68-7 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 328-68-7
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD00236641
InChI Key WBTHOSZMTIPJLR-UHFFFAOYSA-N
Synonym 3-amino-5-trifluoromethyl benzoic acid, 3-amino-5-trifluoromethyl benzoicacid, 5-amino-3-trifluoromethyl benzoic acid, benzoic acid, 3-amino-5-trifluoromethyl, pubchem1359, acmc-209xwk, ksc495q1r, 3-amino-5-carboxybenzotrifluoride, buttpark 25\01-23, 3-carboxy-5-trifluoromethyl aniline
PubChem CID 609556
IUPAC Name 3-amino-5-(trifluoromethyl)benzoic acid
SMILES NC1=CC(=CC(=C1)C(F)(F)F)C(O)=O

Specifications

Melting Point 142°C to 146°C
Quantity 5 g
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material 3-Amino-5-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only