missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-tert-butylisoxazole, 97%

Catalog No. p-7025196
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2056706 5 g
AAB2056714 25 g
AAB2056722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2056706 Supplier Thermo Scientific Chemicals Supplier No. B2056706
Only null left

CAS: 55809-36-4 | C7H12N2O | 140.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55809-36-4
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00055620
InChI Key GGXGVZJHUKEJHO-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butylisoxazole, 5-tert-butylisoxazol-3-amine, 3-amino-5-t-butylisoxazole, 5-tert-butyl isoxazol-3-amine, 3-isoxazolamine, 5-1,1-dimethylethyl, 3-amino-5-tertbutylisoxazole, 5-tert-butyl-isoxazol-3-ylamine, 3-amino-5-tert-butyl isoxazole, 5-tert-butyl isoxazole-3-ylamine
PubChem CID 171473
IUPAC Name 5-tert-butyl-1,2-oxazol-3-amine
SMILES CC(C)(C)C1=CC(=NO1)N

Specifications

Melting Point 110°C to 113°C
Quantity 5 g
Beilstein 1100990
Formula Weight 140.19
Percent Purity 97%
Chemical Name or Material 3-Amino-5-tert-butylisoxazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only