accessibility menu, dialog, popup

UserName

CAS RN 55809-36-4

IUPAC Name: 5-tert-butyl-1,2-oxazol-3-amine
Synonyms: 5-tert-butyl-1,2-oxazol-3-amine
Molecular Weight (g/mol): 140.19
Molecular Formula: C7H12N2O
InChi Key: GGXGVZJHUKEJHO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(N)=NO1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)