missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-tert-butylisoxazole 97.0+%, TCI America™
$171.34 - $171.34
Chemical Identifiers
| CAS | 55809-36-4 |
|---|---|
| Molecular Formula | C7H12N2O |
| Molecular Weight (g/mol) | 140.186 |
| MDL Number | MFCD00055620 |
| InChI Key | GGXGVZJHUKEJHO-UHFFFAOYSA-N |
| Synonym | 3-amino-5-tert-butylisoxazole, 5-tert-butylisoxazol-3-amine, 3-amino-5-t-butylisoxazole, 5-tert-butyl isoxazol-3-amine, 3-isoxazolamine, 5-1,1-dimethylethyl, 3-amino-5-tertbutylisoxazole, 5-tert-butyl-isoxazol-3-ylamine, 3-amino-5-tert-butyl isoxazole, 5-tert-butyl isoxazole-3-ylamine |
| PubChem CID | 171473 |
| IUPAC Name | 5-tert-butyl-1,2-oxazol-3-amine |
| SMILES | CC(C)(C)C1=CC(=NO1)N |
Chemical Identifiers
| 55809-36-4 | |
| 140.186 | |
| GGXGVZJHUKEJHO-UHFFFAOYSA-N | |
| 171473 | |
| CC(C)(C)C1=CC(=NO1)N |
| C7H12N2O | |
| MFCD00055620 | |
| 3-amino-5-tert-butylisoxazole, 5-tert-butylisoxazol-3-amine, 3-amino-5-t-butylisoxazole, 5-tert-butyl isoxazol-3-amine, 3-isoxazolamine, 5-1,1-dimethylethyl, 3-amino-5-tertbutylisoxazole, 5-tert-butyl-isoxazol-3-ylamine, 3-amino-5-tert-butyl isoxazole, 5-tert-butyl isoxazole-3-ylamine | |
| 5-tert-butyl-1,2-oxazol-3-amine |
Specifications
| 55809-36-4 | |
| White-Yellow | |
| MFCD00055620 | |
| 3-amino-5-tert-butylisoxazole, 5-tert-butylisoxazol-3-amine, 3-amino-5-t-butylisoxazole, 5-tert-butyl isoxazol-3-amine, 3-isoxazolamine, 5-1,1-dimethylethyl, 3-amino-5-tertbutylisoxazole, 5-tert-butyl-isoxazol-3-ylamine, 3-amino-5-tert-butyl isoxazole, 5-tert-butyl isoxazole-3-ylamine | |
| CC(C)(C)C1=CC(=NO1)N | |
| 140.186 | |
| 140.19 | |
| Crystalline Powder |
| 112°C | |
| C7H12N2O | |
| 5 g | |
| GGXGVZJHUKEJHO-UHFFFAOYSA-N | |
| 5-tert-butyl-1,2-oxazol-3-amine | |
| 171473 | |
| ≥97.0% (GC,T) | |
| 3-Amino-5-tert-butylisoxazole |
Safety and Handling
EINECSNumber : (9)-2175
TSCA : No