missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-tert-butylisoxazole 97.0+%, TCI America™

Catalog No. A23415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23415G Supplier TCI America Supplier No. A23415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55809-36-4
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00055620
InChI Key GGXGVZJHUKEJHO-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butylisoxazole, 5-tert-butylisoxazol-3-amine, 3-amino-5-t-butylisoxazole, 5-tert-butyl isoxazol-3-amine, 3-isoxazolamine, 5-1,1-dimethylethyl, 3-amino-5-tertbutylisoxazole, 5-tert-butyl-isoxazol-3-ylamine, 3-amino-5-tert-butyl isoxazole, 5-tert-butyl isoxazole-3-ylamine
PubChem CID 171473
IUPAC Name 5-tert-butyl-1,2-oxazol-3-amine
SMILES CC(C)(C)C1=CC(=NO1)N

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 5 g
Formula Weight 140.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-tert-butylisoxazole
EINECSNumber (9)-2175
TSCA No