missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-nitrobenzoic acid, 98%

Catalog No. AAA1250806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1250806 5 g
AAA1250814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1250806 Supplier Thermo Scientific Chemicals Supplier No. A1250806

CAS: 618-84-8 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-84-8
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00017025
InChI Key ZNVHAQRPXAQKRU-UHFFFAOYSA-N
Synonym 3-amino-5-nitrobenzoic acid, 3-amino-5-nitro-benzoic acid, benzoic acid, 3-amino-5-nitro, 3-amino-5-nitrobenzoicacid, 5-amino-3-nitrobenzoic acid, acmc-20ana2, 3-nitro-5-aminobenzoic acid, 5-nitro-3-aminobenzoic acid, benzoic acid,3-amino-5-nitro, 3-amino-5-nitrobenzoic acid 1g
PubChem CID 239342
SMILES C1=C(C=C(C=C1N)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 214°C to 218°C
Quantity 5 g
Beilstein 1457935
Sensitivity Hygroscopic
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 3-Amino-5-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only