missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methylthio-1H-1,2,4-triazole 97.0+%, TCI America™

Catalog No. A14625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14625G 5 g
A146225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14625G Supplier TCI America Supplier No. A14625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 45534-08-5
Molecular Formula C3H6N4S
Molecular Weight (g/mol) 130.17
MDL Number MFCD00005232
InChI Key XGWWZKBCQLBJNH-UHFFFAOYSA-N
Synonym 3-amino-5-methylthio-1h-1,2,4-triazole, 1h-1,2,4-triazol-3-amine, 5-methylthio, 5-methylthio-1h-1,2,4-triazol-3-amine, 3-amino-5-methylthio-1,2,4-triazole, 5-methylsulfanyl-4h-1,2,4-triazol-3-amine, 3-methylthio-1h-1,2,4-triazol-5-amine, 3-methylsulfanyl-1h-1,2,4-triazol-5-amine, 5-methylsulfanyl-1h-1,2,4-triazol-3-amine, 5-amino-3-methylthio-1h-1,2,4-triazole, 5-methylthio-4h-1,2,4-triazol-3-amine
PubChem CID 78471
IUPAC Name 3-(methylsulfanyl)-1H-1,2,4-triazol-5-amine
SMILES CSC1=NNC(N)=N1

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 130.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-methylthio-1H-1,2,4-triazole