accessibility menu, dialog, popup

UserName

CAS RN 45534-08-5

IUPAC Name: 3-(methylsulfanyl)-1H-1,2,4-triazol-5-amine
Synonyms: 5-(methylsulfanyl)-2H-1,2,4-triazol-3-amine
Molecular Weight (g/mol): 130.17
Molecular Formula: C3H6N4S
InChi Key: XGWWZKBCQLBJNH-UHFFFAOYSA-N
SMILES: CSC1=NNC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results