missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methyl-1H-pyrazole, 97%

CAS: 31230-17-8 | C4H7N3 | 97.121 g/mol

Supplier:  Thermo Scientific Chemicals A1164214

 View more versions of this product

Catalog No. AAA1164214


Only null left
Explore available promotions

Description

Description

3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

Solubility
Soluble in Dichloromethane and Methanol.

Notes
Air Sensitive. Store at -20°C. Store under dry inert gas away from oxidizing agents and air.
Specifications

Specifications

3-Amino-5-methyl-1H-pyrazole
42°C to 47°C
>110°C (230°F)
MFCD00075180
1904
3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
FYTLHYRDGXRYEY-UHFFFAOYSA-N
5-methyl-1H-pyrazol-3-amine
93146
97%
31230-17-8
212°C to 213°C (14 mmHg)
C4H7N3
25 g
Air Sensitive
Soluble in Dichloromethane and Methanol.
CC1=CC(=NN1)N
97.121
97.12

Chemical Identifiers

31230-17-8
97.121
FYTLHYRDGXRYEY-UHFFFAOYSA-N
93146
CC1=CC(=NN1)N
C4H7N3
MFCD00075180
3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
5-methyl-1H-pyrazol-3-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only