missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-hydroxypyrazole 98.0+%, TCI America™

Catalog No. p-7117537
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15895G 5 g
A158925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15895G Supplier TCI America Supplier No. A15895G
Only null left

Chemical Identifiers

CAS 6126-22-3
Molecular Formula C3H5N3O
Molecular Weight (g/mol) 99.093
MDL Number MFCD00022384
InChI Key QZBGOTVBHYKUDS-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxypyrazole, 3-amino-1h-pyrazol-5-ol, 5-amino-1h-pyrazol-3-ol, 5-amino-1h-pyrazol-3 2h-one, 3-amino-1,2-dihydropyrazol-5-one, 5-aminopyrazol-3-ol, 5-amino-2h-pyrazol-3-ol, 3-pyrazolin-5-one, 3-amino, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 1h-pyrazol-5-ol, 3-amino
PubChem CID 96221
IUPAC Name 5-amino-1,2-dihydropyrazol-3-one
SMILES C1=C(NNC1=O)N

Specifications

Quantity 5 g
Formula Weight 99.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-hydroxypyrazole