accessibility menu, dialog, popup

UserName

NH2NNHO

CAS RN 6126-22-3

IUPAC Name: 3-amino-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 5-amino-2,4-dihydropyrazol-3-one
Molecular Weight (g/mol): 99.09
Molecular Formula: C3H5N3O
InChi Key: UELIBMUMKLRTKJ-UHFFFAOYSA-N
SMILES: NC1=NNC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)